Assay Details
Target Gene Details
Entrez Gene ID: | 80212 |
Gene Name: | coiled-coil domain containing 92 |
Gene Aliases: |
- |
Location: |
Chr.12:123936409-123972985 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC92 | NM_001304957.1 | 5 | 1429 | NP_001291886.1 |
| NM_001304958.1 | 6 | 1536 | NP_001291887.1 | |
| NM_001304959.1 | 5 | 1178 | NP_001291888.1 | |
| NM_001304960.1 | 7 | 1347 | NP_001291889.1 | |
| NM_001304961.1 | 4 | 1346 | NP_001291890.1 | |
| NM_025140.2 | 5 | 1439 | NP_079416.1 | |
| XM_005253624.2 | 5 | 1183 | XP_005253681.1 | |
| XM_017019984.1 | 5 | 1997 | XP_016875473.1 | |
| AB015292.1 | 4 | 959 | BAB82436.1 | |
| AK026124.1 | 5 | 1070 | BAB15366.1 | |
| AK054680.1 | 3 | 2706 | ||
| AK124358.1 | 1 | 2528 | ||
| AK125866.1 | 3 | 2957 | ||
| AK222661.1 | 5 | 1027 | BAD96381.1 | |
| BC017186.2 | 5 | 1062 | AAH17186.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522321 | Chr.12:123899403 - 123940089 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map