Assay Details
Target Gene Details
Entrez Gene ID: | 7165 |
Gene Name: | tumor protein D52 like 2 |
Gene Aliases: |
D54, TPD54 |
Location: |
Chr.20:63865228-63891545 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPD52L2 | NM_001243891.1 | 1 | 44 | NP_001230820.1 |
| NM_001243892.1 | 1 | 44 | NP_001230821.1 | |
| NM_001243894.1 | 1 | 44 | NP_001230823.1 | |
| NM_001243895.1 | 1 | 44 | NP_001230824.1 | |
| NM_003288.3 | 1 | 44 | NP_003279.2 | |
| NM_199359.2 | 1 | 44 | NP_955391.1 | |
| NM_199360.2 | 1 | 44 | NP_955392.1 | |
| NM_199361.2 | 1 | 44 | NP_955393.1 | |
| NM_199362.2 | 1 | 44 | NP_955394.1 | |
| NM_199363.2 | 1 | 44 | NP_955395.1 | |
| NR_045090.1 | 1 | 44 | ||
| XM_011529034.2 | 1 | 44 | XP_011527336.1 | |
| XM_011529035.2 | 1 | 44 | XP_011527337.1 | |
| XM_017028038.1 | XP_016883527.1 | |||
| AF004430.1 | AAC98478.1 | |||
| AK055068.1 | ||||
| AK297064.1 | ||||
| AK298366.1 | ||||
| AK314501.1 | ||||
| AU124850.1 | ||||
| BM473873.1 | ||||
| BM833522.1 | ||||
| BQ948007.1 | ||||
| DA871584.1 | 1 | 44 | ||
| DC362382.1 | ||||
| DC428780.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509784 | Chr.20:63824382 - 63983325 on Build GRCh38 | Insertion |
|
| nsv428381 | Chr.20:63583375 - 64334167 on Build GRCh38 | Gain+Loss |
|
| nsv469773 | Chr.20:63845428 - 64039312 on Build GRCh38 | Gain |
|
| nsv828804 | Chr.20:63809574 - 63878524 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map