Assay Details
Target Gene Details
Entrez Gene ID: | 79956 |
Gene Name: | endoplasmic reticulum metallopeptidase 1 |
Gene Aliases: |
FXNA, KIAA1815, bA207C16.3 |
Location: |
Chr.9:5764061-5867091 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERMP1 | AK127218.1 | 16 | 3063 |
Target Gene Details
Entrez Gene ID: | 57589 |
Gene Name: | RIC1 homolog, RAB6A GEF complex partner 1 |
Gene Aliases: |
CIP150, KIAA1432, bA207C16.1 |
Location: |
Chr.9:5629030-5778633 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 22 - Intron 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RIC1 | NM_001135920.2 | NP_001129392.2 | ||
| NM_001206557.1 | NP_001193486.1 | |||
| NM_020829.3 | NP_065880.2 | |||
| XM_005251523.3 | XP_005251580.1 | |||
| XM_011517967.2 | XP_011516269.1 | |||
| XM_011517968.2 | XP_011516270.1 | |||
| XM_017014934.1 | XP_016870423.1 | |||
| XM_017014935.1 | XP_016870424.1 | |||
| XM_017014936.1 | XP_016870425.1 | |||
| AB037853.2 | BAA92670.2 | |||
| AB205401.1 | BAE16982.1 | |||
| AK022642.1 | BAB14150.1 | |||
| AK074150.1 | BAB84976.1 | |||
| AK124394.1 | ||||
| AL136875.1 | CAB66809.1 | |||
| BC136616.1 | ||||
| BC144296.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3573095 | Chr.9:5721091 - 5789971 on Build GRCh38 | Loss |
|
| nsv1029665 | Chr.9:5687332 - 6775107 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map