Assay Details
Target Gene Details
Entrez Gene ID: | 100528018 |
Gene Name: | ARL2-SNX15 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.11:65014113-65040572 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARL2-SNX15 | NR_037650.1 | 11 | 1835 | |
| AK096447.1 | 11 | 1769 |
Target Gene Details
Entrez Gene ID: | 29907 |
Gene Name: | sorting nexin 15 |
Gene Aliases: |
HSAF001435 |
Location: |
Chr.11:65027408-65040572 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNX15 | NM_013306.4 | 8 | 1311 | NP_037438.2 |
| NM_147777.3 | 7 | 1053 | NP_680086.2 | |
| AB209439.1 | 7 | 4089 | BAD92676.1 | |
| AF001435.1 | 8 | 1240 | AAB81205.1 | |
| AF175267.2 | 8 | 1220 | AAF89955.1 | |
| BC009897.2 | 8 | 1229 | AAH09897.1 | |
| BC012767.2 | 8 | 1281 | AAH12767.1 | |
| BC014520.2 | 8 | 1281 | AAH14520.1 | |
| BE547440.1 | 2 | 321 | ||
| BE884806.1 | 2 | 387 | ||
| BX444780.2 | 1 | 614 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526181 | Chr.11:64870569 - 65489858 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map