Assay Details
Target Gene Details
Entrez Gene ID: | 128822 |
Gene Name: | cystatin 9 |
Gene Aliases: |
CLM, CTES7A |
Location: |
Chr.20:23602410-23605973 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CST9 | NM_001008693.2 | 2 | 570 | NP_001008693.2 |
| AF494536.1 | 2 | 473 | AAM18055.1 | |
| BC137302.1 | 2 | 570 | ||
| BC137303.1 | 2 | 570 |
More Information
Set Membership: |
|

Genomic Map