Assay Details
Target Gene Details
Entrez Gene ID: | 1211 |
Gene Name: | clathrin light chain A |
Gene Aliases: |
LCA |
Location: |
Chr.9:36190856-36212062 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLTA | NM_001076677.2 | 1 | 54 | NP_001070145.1 |
| NM_001184760.1 | 1 | 54 | NP_001171689.1 | |
| NM_001184761.1 | 1 | 54 | NP_001171690.1 | |
| NM_001184762.1 | 1 | 54 | NP_001171691.1 | |
| NM_001311203.1 | 1 | 54 | NP_001298132.1 | |
| NM_001311204.1 | 1 | 54 | NP_001298133.1 | |
| NM_001311205.1 | 1 | 54 | NP_001298134.1 | |
| NM_001311206.1 | 1 | 54 | NP_001298135.1 | |
| NM_001833.3 | 1 | 54 | NP_001824.1 | |
| NM_007096.3 | 1 | 54 | NP_009027.1 | |
| NR_132349.1 | 1 | 54 | ||
| XM_017014257.1 | XP_016869746.1 | |||
| XM_017014258.1 | XP_016869747.1 | |||
| AK291078.1 | ||||
| AK295692.1 | ||||
| BC009201.2 | AAH09201.1 | |||
| BI090227.1 | ||||
| CN300759.1 | ||||
| DT220470.1 | 1 | 54 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1017711 | Chr.9:36073424 - 36548245 on Build GRCh38 | Gain |
|
| esv34080 | Chr.9:36005887 - 36453179 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map