Assay Details
Target Gene Details
Entrez Gene ID: | 10111 |
Gene Name: | RAD50 double strand break repair protein |
Gene Aliases: |
NBSLD, RAD502, hRad50 |
Location: |
Chr.5:132556924-132644621 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD50 | NM_005732.3 | 1 | 302 | NP_005723.2 |
| AF057299.1 | 1 | 223 | AAD50325.1 | |
| AF057300.1 | 1 | 223 | AAD50326.1 | |
| AK290597.1 | 1 | 244 | ||
| AK307439.1 | ||||
| BC062603.1 | 1 | 219 | AAH62603.1 | |
| BC073850.1 | 1 | 219 | AAH73850.1 | |
| BC108282.1 | 1 | 239 | AAI08283.1 | |
| BC136436.1 | 1 | 194 | ||
| BC140005.1 | 1 | 296 | ||
| DB112912.1 | 1 | 303 | ||
| DQ655928.2 | ||||
| U63139.1 | 1 | 288 | AAB07119.1 |
More Information
Set Membership: |
|

Genomic Map