Assay Details
Target Gene Details
Entrez Gene ID: | 4354 |
Gene Name: | membrane palmitoylated protein 1 |
Gene Aliases: |
AAG12, DXS552E, EMP55, MRG1, PEMP |
Location: |
Chr.X:154778684-154805527 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPP1 | NM_001166460.1 | 12 | 1586 | NP_001159932.1 |
| NM_001166461.1 | 12 | 1577 | NP_001159933.1 | |
| NM_001166462.1 | 13 | 1786 | NP_001159934.1 | |
| NM_002436.3 | 12 | 1637 | NP_002427.1 | |
| XM_011531167.1 | 11 | 1550 | XP_011529469.1 | |
| XM_011531169.1 | 8 | 1209 | XP_011529471.1 | |
| AK093702.1 | 3 | 1969 | ||
| AK223396.1 | 12 | 1617 | BAD97116.1 | |
| AK290246.1 | ||||
| AK303111.1 | 12 | 1452 | ||
| AK304538.1 | 12 | 1544 | ||
| AK315957.1 | 13 | 1635 | ||
| BC002392.2 | 12 | 1513 | AAH02392.1 | |
| M64925.1 | 12 | 1586 | AAA60059.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map