Assay Details
Target Gene Details
Entrez Gene ID: | 1869 |
Gene Name: | E2F transcription factor 1 |
Gene Aliases: |
E2F-1, RBAP1, RBBP3, RBP3 |
Location: |
Chr.20:33675486-33686404 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| E2F1 | NM_005225.2 | NP_005216.1 | ||
| BC050369.2 | AAH50369.1 | |||
| BC058902.1 | AAH58902.1 | |||
| M96577.1 | AAA35782.1 | |||
| S49592.1 | AAB24289.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv954024 | Chr.20:33638795 - 33698694 on Build GRCh38 | Deletion |
|
| nsv1065189 | Chr.20:33597378 - 33807579 on Build GRCh38 | Loss |
|
| nsv833961 | Chr.20:33605380 - 33836992 on Build GRCh38 | Loss |
|
| nsv833960 | Chr.20:33569059 - 33688060 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map