Assay Details
Target Gene Details
Entrez Gene ID: | 2891 |
Gene Name: | glutamate ionotropic receptor AMPA type subunit 2 |
Gene Aliases: |
GLUR2, GLURB, GluA2, GluR-K2, HBGR2 |
Location: |
Chr.4:157205099-157370583 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIA2 | NM_000826.3 | 1 | 492 | NP_000817.2 |
| NM_001083619.1 | 1 | 492 | NP_001077088.1 | |
| NM_001083620.1 | NP_001077089.1 | |||
| XM_017008114.1 | XP_016863603.1 | |||
| XM_017008115.1 | XP_016863604.1 | |||
| XM_017008116.1 | XP_016863605.1 | |||
| AB209567.1 | 1 | 322 | BAD92804.1 | |
| AK294458.1 | ||||
| AK309067.1 | ||||
| AL708924.1 | 1 | 425 | ||
| AL832605.3 | 1 | 352 | ||
| BC010574.1 | 1 | 312 | AAH10574.1 | |
| BC028736.1 | AAH28736.2 | |||
| BP202382.1 | 1 | 383 | ||
| BP202686.1 | ||||
| DA228330.1 | 1 | 461 | ||
| DA810082.1 | 1 | 494 | ||
| L20814.1 | 1 | 193 | AAA58631.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830123 | Chr.4:157214177 - 157362103 on Build GRCh38 | Gain |
|
| nsv1018387 | Chr.4:157038779 - 157843038 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map