Assay Details
Target Gene Details
Entrez Gene ID: | 57732 |
Gene Name: | zinc finger FYVE-type containing 28 |
Gene Aliases: |
LST2, LYST2 |
Location: |
Chr.4:2269585-2418698 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFYVE28 | NM_001172656.1 | NP_001166127.1 | ||
| NM_001172659.1 | NP_001166130.1 | |||
| NM_020972.2 | NP_066023.2 | |||
| XM_006713900.3 | XP_006713963.1 | |||
| XM_006713902.3 | XP_006713965.1 | |||
| XM_011513540.2 | XP_011511842.1 | |||
| XM_011513541.2 | XP_011511843.1 | |||
| XM_017008502.1 | XP_016863991.1 | |||
| XM_017008503.1 | XP_016863992.1 | |||
| XM_017008504.1 | XP_016863993.1 | |||
| XM_017008505.1 | XP_016863994.1 | |||
| XM_017008510.1 | XP_016863999.1 | |||
| AB046863.1 | ||||
| AK126692.1 | ||||
| AK293775.1 | ||||
| AK294710.1 | ||||
| AK294874.1 | ||||
| AK316484.1 | ||||
| BC041866.1 | AAH41866.1 | |||
| BC137309.1 | ||||
| BC137310.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508990 | Chr.4:2246239 - 2324327 on Build GRCh38 | Insertion |
|
| nsv4200 | Chr.4:2252081 - 2286377 on Build GRCh38 | Insertion |
|
| nsv1009347 | Chr.4:2200935 - 2516063 on Build GRCh38 | Gain |
|
| esv2726835 | Chr.4:2019561 - 2959568 on Build GRCh38 | Deletion |
|
| nsv527189 | Chr.4:2243138 - 2276689 on Build GRCh38 | Loss |
|
| nsv829838 | Chr.4:2155167 - 2349950 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map