Assay Details
Target Gene Details
Entrez Gene ID: | 54520 |
Gene Name: | coiled-coil domain containing 93 |
Gene Aliases: |
- |
Location: |
Chr.2:117915478-118014163 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC93 | NM_019044.4 | 24 | 6717 | NP_061917.3 |
| XM_011511359.1 | 24 | 6714 | XP_011509661.1 | |
| XM_011511361.1 | 23 | 6471 | XP_011509663.1 | |
| XM_011511362.1 | 23 | 6585 | XP_011509664.1 | |
| AB209993.1 | 24 | 6636 | ||
| AF070582.1 | 1 | 1625 | ||
| AK124683.1 | 1 | 2761 | ||
| AL050001.1 | 1 | 1017 | ||
| AY726577.1 | 1 | 2681 | ||
| BU621185.1 | 1 | 230 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv999434 | Chr.2:117074067 - 117975257 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map