Assay Details
Target Gene Details
Entrez Gene ID: | 5949 |
Gene Name: | retinol binding protein 3 |
Gene Aliases: |
D10S64, D10S65, D10S66, IRBP, RBPI, RP66 |
Location: |
Chr.10:47348371-47357875 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBP3 | NM_002900.2 | 1 | 2454 | NP_002891.1 |
| AB593121.1 | 1 | 2446 | ||
| BC039844.1 | 1 | 2444 | AAH39844.1 | |
| J03912.1 | 1 | 2337 | AAA59188.1 | |
| M22453.1 | 1 | 2454 | AAA36126.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831861 | Chr.10:47306097 - 47471541 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map