Assay Details
Target Gene Details
Entrez Gene ID: | 115557 |
Gene Name: | Rho guanine nucleotide exchange factor 25 |
Gene Aliases: |
GEFT, p63RhoGEF |
Location: |
Chr.12:57610180-57617245 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGEF25 | NM_001111270.2 | NP_001104740.1 | ||
| NM_182947.3 | NP_891992.2 | |||
| NR_046223.1 | ||||
| AB370196.1 | ||||
| AF487514.2 | AAO49463.2 | |||
| BC012860.1 | AAH12860.1 | |||
| BC047559.1 | AAH47559.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv559034 | Chr.12:57466149 - 57618450 on Build GRCh38 | Loss |
|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map