Assay Details
Target Gene Details
Entrez Gene ID: | 4287 |
Gene Name: | ataxin 3 |
Gene Aliases: |
AT3, ATX3, JOS, MJD, MJD1, SCA3 |
Location: |
Chr.14:92058552-92106621 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv565519 | Chr.14:92033004 - 92089711 on Build GRCh38 | Loss |
|
| dgv1956n100 | Chr.14:92056253 - 92104511 on Build GRCh38 | Gain |
|
| nsv1045760 | Chr.14:92030812 - 92090267 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map