Assay Details
Target Gene Details
Entrez Gene ID: | 155 |
Gene Name: | adrenoceptor beta 3 |
Gene Aliases: |
BETA3AR |
Location: |
Chr.8:37962995-37966666 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADRB3 | NM_000025.2 | 2 | 1969 | NP_000016.1 |
| X70811.1 | 2 | 1897 | CAA50141.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv610947 | Chr.8:37935047 - 38054626 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map