Assay Details
Target Gene Details
Entrez Gene ID: | 388341 |
Gene Name: | leucine rich repeat containing 75A |
Gene Aliases: |
C17orf76, FAM211A |
Location: |
Chr.17:16442005-16492191 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC75A | NM_001113567.2 | NP_001107039.1 | ||
| NM_207387.3 | NP_997270.2 | |||
| XM_011523845.2 | XP_011522147.1 | |||
| XM_017024619.1 | XP_016880108.1 | |||
| XM_017024620.1 | XP_016880109.1 | |||
| AI298465.1 | ||||
| AK093015.1 | BAC04020.1 | |||
| AL833245.1 | 3 | 2672 | ||
| AL833260.1 | 3 | 2672 |
Target Gene Details
Entrez Gene ID: | 125144 |
Gene Name: | LRRC75A antisense RNA 1 |
Gene Aliases: |
C17orf45, C17orf76-AS1, FAM211A-AS1, NCRNA00188, TSAP19 |
Location: |
Chr.17:16438987-16470648 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC75A-AS1 | NR_027158.1 | |||
| NR_027159.1 | ||||
| NR_027160.1 | ||||
| NR_027161.1 | ||||
| NR_027162.1 | ||||
| NR_027163.1 | ||||
| NR_027164.1 | ||||
| NR_027165.1 | ||||
| NR_027166.1 | ||||
| NR_027167.1 | ||||
| NR_027168.2 | ||||
| NR_027169.1 | ||||
| NR_027170.1 | ||||
| NR_027171.1 | ||||
| NR_027172.2 | ||||
| NR_027173.1 | ||||
| NR_027174.1 | ||||
| NR_027175.1 | ||||
| NR_027176.1 | ||||
| NR_027177.1 | ||||
| NR_027178.1 | ||||
| NR_027667.1 | ||||
| NR_045021.1 | ||||
| NR_045022.1 | ||||
| NR_045023.1 | ||||
| NR_045025.1 | ||||
| NR_045027.1 | ||||
| NR_045028.1 | ||||
| NR_045029.1 | ||||
| AJ012499.1 | ||||
| AK098643.1 | ||||
| AK308482.1 | ||||
| AK309961.1 | ||||
| BC012396.1 | AAH12396.1 | |||
| BC019833.1 | AAH19833.1 | |||
| BC027986.1 | AAH27986.1 | |||
| BC040159.1 | AAH40159.1 | |||
| BP377665.1 | ||||
| BX538291.1 | CAD98086.1 | |||
| DA131201.1 | ||||
| DA193585.1 | ||||
| DA383448.1 | ||||
| DA485648.1 | ||||
| DA494627.1 | ||||
| DA769571.1 | ||||
| DA793334.1 | ||||
| DA802938.1 | ||||
| DA857697.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1063764 | Chr.17:16268335 - 16759600 on Build GRCh38 | Loss |
|
| esv2715702 | Chr.17:15750514 - 16668268 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map