Assay Details
Target Gene Details
Entrez Gene ID: | 121275 |
Gene Name: | olfactory receptor family 10 subfamily AD member 1 |
Gene Aliases: |
OR10AD1P, OR12-1 |
Location: |
Chr.12:48202339-48203292 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR10AD1 | NM_001004134.1 | 1 | 728 | NP_001004134.1 |
| BC136757.1 | 1 | 823 | ||
| BC136758.1 | 1 | 823 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759898 | Chr.12:48035919 - 48431716 on Build GRCh38 | Gain+Loss |
|
| esv33956 | Chr.12:48171860 - 48251778 on Build GRCh38 | Loss |
|
| esv3629429 | Chr.12:48142951 - 48278499 on Build GRCh38 | Gain |
|
| nsv8971 | Chr.12:48142743 - 48364718 on Build GRCh38 | Gain+Loss |
|
| esv34439 | Chr.12:48147974 - 48385779 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map