Assay Details
Target Gene Details
Entrez Gene ID: | 92591 |
Gene Name: | ankyrin repeat and SOCS box containing 16 |
Gene Aliases: |
- |
Location: |
Chr.17:44170706-44179083 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB16 | NM_080863.4 | NP_543139.4 | ||
| AK054727.1 | BAB70800.1 | |||
| AK314595.1 | ||||
| AY557346.1 | AAS67920.1 | |||
| BC075088.2 | AAH75088.1 | |||
| BC118584.1 | ||||
| BC118585.1 |
Target Gene Details
Entrez Gene ID: | 339201 |
Gene Name: | ASB16 antisense RNA 1 |
Gene Aliases: |
C17orf65 |
Location: |
Chr.17:44175973-44186717 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB16-AS1 | NR_049729.1 | 4 | 470 | |
| NR_049730.1 | ||||
| AK055685.1 | 4 | 424 | ||
| CB151608.1 | ||||
| CN367999.1 | ||||
| DA567080.1 | 3 | 306 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833455 | Chr.17:44108553 - 44297300 on Build GRCh38 | Loss |
|
| dgv529n67 | Chr.17:44170904 - 44246444 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map