Assay Details
Target Gene Details
Entrez Gene ID: | 1493 |
Gene Name: | cytotoxic T-lymphocyte associated protein 4 |
Gene Aliases: |
ALPS5, CD, CD152, CELIAC3, CTLA-4, GRD4, GSE, IDDM12 |
Location: |
Chr.2:203867788-203873960 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTLA4 | NM_001037631.2 | 3 | 1510 | NP_001032720.1 |
| NM_005214.4 | 4 | 1620 | NP_005205.2 | |
| AF414120.1 | 4 | 1620 | AAL07473.1 | |
| BG058720.1 | 1 | 367 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001799 | Chr.2:202908732 - 205615330 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map