Assay Details
Target Gene Details
Entrez Gene ID: | 3268 |
Gene Name: | ArfGAP with FG repeats 2 |
Gene Aliases: |
HRBL, RABR |
Location: |
Chr.7:100539211-100568220 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGFG2 | NM_006076.4 | 12 | 3040 | NP_006067.3 |
| XM_005250306.2 | 13 | 3059 | XP_005250363.1 | |
| XM_017012112.1 | 12 | 2981 | XP_016867601.1 | |
| XM_017012114.1 | 11 | 2739 | XP_016867603.1 | |
| AK090744.1 | 1 | 807 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion |
|
| nsv951386 | Chr.7:100566478 - 100682977 on Build GRCh38 | Deletion |
|
| nsv831072 | Chr.7:100564880 - 100765212 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map