Assay Details
Target Gene Details
Entrez Gene ID: | 4609 |
Gene Name: | v-myc avian myelocytomatosis viral oncogene homolog |
Gene Aliases: |
MRTL, MYCC, bHLHe39, c-Myc |
Location: |
Chr.8:127736069-127741434 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYC | NM_002467.4 | 3 | 1652 | NP_002458.2 |
| AK303921.1 | 4 | 1434 | ||
| AK312883.1 | 3 | 1485 | ||
| BC000141.1 | 3 | 1463 | ||
| BC000917.2 | 3 | 1479 | ||
| BC058901.1 | 3 | 1165 | ||
| BT019768.1 | 2 | 1082 | AAV38573.1 | |
| K02276.1 | 3 | 1502 | AAA36340.1 | |
| KC782559.1 | 2 | 355 | ||
| V00568.1 | 3 | 1640 | CAA23831.1 | |
| X66258.1 | CAA46984.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516713 | Chr.8:127727525 - 127755908 on Build GRCh38 | Gain+Loss |
|
| nsv831458 | Chr.8:127702603 - 127875740 on Build GRCh38 | Loss |
|
| esv2752208 | Chr.8:127632573 - 127895715 on Build GRCh38 | Gain |
|
| esv3618781 | Chr.8:127738039 - 127746724 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map