Assay Details
Target Gene Details
Entrez Gene ID: | 23253 |
Gene Name: | ankyrin repeat domain 12 |
Gene Aliases: |
ANCO-2, ANCO1, GAC-1, Nbla00144 |
Location: |
Chr.18:9136753-9285985 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD12 | NM_001083625.2 | 1 | 137 | NP_001077094.1 |
| NM_015208.4 | 1 | 137 | NP_056023.3 | |
| XM_005258093.3 | 1 | 123 | XP_005258150.1 | |
| XM_005258095.2 | 1 | 109 | XP_005258152.1 | |
| XM_005258096.2 | 1 | 113 | XP_005258153.1 | |
| XM_011525638.2 | 1 | 109 | XP_011523940.1 | |
| XM_011525639.2 | 1 | 95 | XP_011523941.1 | |
| XM_017025661.1 | 1 | 107 | XP_016881150.1 | |
| XM_017025663.1 | 1 | 91 | XP_016881152.1 | |
| XM_017025664.1 | 1 | 91 | XP_016881153.1 | |
| XM_017025667.1 | XP_016881156.1 | |||
| XM_017025668.1 | XP_016881157.1 | |||
| AK024808.1 | 1 | 87 | BAB15014.1 | |
| AL834204.1 | 1 | 65 | CAH56382.1 | |
| AY373757.1 | 1 | 87 | AAR25662.1 | |
| BC057225.1 | 1 | 86 | AAH57225.1 | |
| BC070234.1 | 1 | 107 | AAH70234.1 | |
| BP230422.1 | 1 | 137 | ||
| BP255733.1 | 1 | 130 |

Genomic Map