Assay Details
Target Gene Details
Entrez Gene ID: | 257236 |
Gene Name: | coiled-coil domain containing 96 |
Gene Aliases: |
- |
Location: |
Chr.4:7040849-7043001 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC96 | NM_153376.2 | 1 | 514 | NP_699207.1 |
| AK075056.1 | 1 | 452 | BAC11373.1 | |
| BC105055.1 | 1 | 510 | AAI05056.1 | |
| BC112274.1 | 1 | 510 | AAI12275.1 | |
| DB045256.1 | 1 | 514 |
Target Gene Details
Entrez Gene ID: | 100129931 |
Gene Name: | uncharacterized LOC100129931 |
Gene Aliases: |
- |
Location: |
Chr.4:7030554-7046231 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100129931 | NR_033828.1 | |||
| AK126065.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428436 | Chr.4:6986421 - 7268174 on Build GRCh38 | Gain |
|
| nsv4217 | Chr.4:7033785 - 7078503 on Build GRCh38 | Deletion |
|
| esv26516 | Chr.4:7042252 - 7044267 on Build GRCh38 | Loss |
|
| esv34033 | Chr.4:6971543 - 7388894 on Build GRCh38 | Loss |
|
| nsv469847 | Chr.4:7001798 - 7166714 on Build GRCh38 | Gain |
|
| nsv593549 | Chr.4:7042493 - 7043930 on Build GRCh38 | Gain+Loss |
|
| nsv593548 | Chr.4:7041372 - 7044218 on Build GRCh38 | Loss |
|

Genomic Map