Assay Details
Target Gene Details
Entrez Gene ID: | 2002 |
Gene Name: | ELK1, ETS transcription factor |
Gene Aliases: |
- |
Location: |
Chr.X:47635520-47650604 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELK1 | NM_001114123.2 | 7 | 1671 | NP_001107595.1 |
| NM_001257168.1 | 6 | 793 | NP_001244097.1 | |
| NM_005229.4 | 6 | 1565 | NP_005220.2 | |
| XM_017029339.1 | 5 | 1386 | XP_016884828.1 | |
| AB016193.1 | 6 | 1449 | BAA36616.1 | |
| AB209047.1 | 5 | 3431 | BAD92284.1 | |
| AF000672.1 | AAD00862.1 | |||
| AK093966.1 | 4 | 614 | BAC04259.1 | |
| BC048296.1 | AAH48296.1 | |||
| BC056150.1 | 5 | 1395 | AAH56150.1 | |
| M25269.1 | 7 | 1664 | AAA52384.1 |
More Information
Set Membership: |
|

Genomic Map