Assay Details
Target Gene Details
Entrez Gene ID: | 147 |
Gene Name: | adrenoceptor alpha 1B |
Gene Aliases: |
ADRA1, ALPHA1BAR |
Location: |
Chr.5:159916733-159989205 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADRA1B | NM_000679.3 | 1 | 191 | NP_000670.1 |
| XM_005265818.3 | 1 | 88 | XP_005265875.1 | |
| XM_005265819.2 | 1 | 88 | XP_005265876.1 | |
| XM_006714821.3 | 1 | 88 | XP_006714884.1 | |
| XM_011534435.1 | 1 | 88 | XP_011532737.1 | |
| XM_011534437.2 | 1 | 88 | XP_011532739.1 | |
| XM_011534438.2 | 1 | 88 | XP_011532740.1 | |
| BC136568.1 | 1 | 112 | ||
| BC136569.1 | 1 | 112 | ||
| L31773.1 | AAB59485.1 | |||
| U03865.1 | 1 | 141 | AAB60352.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830531 | Chr.5:159896337 - 160076045 on Build GRCh38 | Loss |
|
| nsv1015634 | Chr.5:159750943 - 160620377 on Build GRCh38 | Gain |
|
| nsv462500 | Chr.5:159884897 - 159921891 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map