Assay Details
Target Gene Details
Entrez Gene ID: | 5074 |
Gene Name: | pro-apoptotic WT1 regulator |
Gene Aliases: |
PAR4, Par-4 |
Location: |
Chr.12:79587791-79691097 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAWR | NM_002583.2 | 5 | 1065 | NP_002574.2 |
| XM_006719435.2 | 5 | 1041 | XP_006719498.1 | |
| XM_017019377.1 | 5 | 945 | XP_016874866.1 | |
| BC007018.1 | 4 | 911 | AAH07018.1 | |
| CR536549.1 | 4 | 779 | CAG38786.1 | |
| U63809.1 | 5 | 1023 | AAC24947.1 |
More Information
Set Membership: |
|

Genomic Map