Assay Details
Target Gene Details
Entrez Gene ID: | 10847 |
Gene Name: | Snf2-related CREBBP activator protein |
Gene Aliases: |
DOMO1, EAF1, FLHS, SWR1 |
Location: |
Chr.16:30699141-30740129 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRCAP | NM_006662.2 | NP_006653.2 | ||
| AB621816.1 | ||||
| AF143946.1 | AAD39760.1 | |||
| AK023808.1 | ||||
| AK128030.1 | BAC87237.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469724 | Chr.16:30621124 - 30828942 on Build GRCh38 | Loss |
|
| nsv833188 | Chr.16:30621125 - 30828942 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map