Assay Details
Target Gene Details
Entrez Gene ID: | 9679 |
Gene Name: | family with sequence similarity 53 member B |
Gene Aliases: |
KIAA0140, bA12J10.2, smp |
Location: |
Chr.10:124619294-124744361 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM53B | NM_014661.3 | NP_055476.3 | ||
| AK127323.1 | ||||
| D50930.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832013 | Chr.10:124557573 - 124776461 on Build GRCh38 | Loss |
|
| nsv552288 | Chr.10:124605790 - 124621473 on Build GRCh38 | Loss |
|
| nsv521161 | Chr.10:124518637 - 124736254 on Build GRCh38 | Gain+Loss |
|
| nsv7589 | Chr.10:124598575 - 124633578 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map