Assay Details
Target Gene Details
Entrez Gene ID: | 84277 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C30 |
Gene Aliases: |
WBSCR18 |
Location: |
Chr.7:73680918-73683451 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC30 | NM_032317.2 | 1 | 108 | NP_115693.2 |
| AF412025.1 | 1 | 99 | AAM62307.1 | |
| AK058113.1 | 1 | 88 | BAB71671.1 | |
| AK094305.1 | 1 | 108 | ||
| BC005056.1 | 1 | 99 | AAH05056.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831028 | Chr.7:73556313 - 73816595 on Build GRCh38 | Loss |
|
| nsv469841 | Chr.7:73621181 - 73770495 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map