Assay Details
Target Gene Details
Entrez Gene ID: | 123228 |
Gene Name: | SUMO/sentrin peptidase family member, NEDD8 specific |
Gene Aliases: |
DEN1, NEDP1, PRSC2 |
Location: |
Chr.15:72114258-72143688 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SENP8 | NM_001166340.1 | 2 | 697 | NP_001159812.1 |
| NM_001172109.1 | 3 | 851 | NP_001165580.1 | |
| NM_001172110.1 | 3 | 693 | NP_001165581.1 | |
| NM_001172111.1 | 3 | 710 | NP_001165582.1 | |
| NM_145204.3 | 2 | 586 | NP_660205.3 | |
| XM_005254157.3 | 2 | 466 | XP_005254214.1 | |
| XM_011521215.2 | 3 | 984 | XP_011519517.1 | |
| XM_011521217.2 | 2 | 574 | XP_011519519.1 | |
| AF308450.1 | 2 | 490 | AAL06294.1 | |
| AK097557.1 | 2 | 697 | ||
| AK292298.1 | 2 | 588 | ||
| AY008293.1 | 2 | 550 | AAG21828.1 | |
| BC031411.1 | 2 | 463 | AAH31411.1 | |
| CN367895.1 | ||||
| DB033241.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833051 | Chr.15:72023130 - 72216088 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map