Assay Details
Target Gene Details
Entrez Gene ID: | 150353 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member B7 |
Gene Aliases: |
DJ5, HSC3 |
Location: |
Chr.22:40859549-40862126 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJB7 | NM_145174.1 | 1 | 113 | NP_660157.1 |
| AF085232.1 | 1 | 113 | AAL40391.1 | |
| BC112135.1 | 1 | 72 | AAI12136.1 | |
| CR456468.1 | CAG30354.1 |
Target Gene Details
Entrez Gene ID: | 63929 |
Gene Name: | X-prolyl aminopeptidase 3 |
Gene Aliases: |
APP3, ICP55, NPHPL1 |
Location: |
Chr.22:40857081-40932819 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XPNPEP3 | NM_001204827.1 | 3 | 678 | NP_001191756.1 |
| NM_022098.3 | NP_071381.1 | |||
| AK301635.1 | ||||
| AK313770.1 | ||||
| AL365514.1 | CAB97210.1 | |||
| AL834310.1 | CAD38980.1 | |||
| BC001208.1 | AAH01208.1 | |||
| BC001681.2 | AAH01681.1 | |||
| BC004989.1 | AAH04989.1 | |||
| BP346902.1 | ||||
| BX648018.1 | ||||
| CR456442.1 | CAG30328.1 | |||
| CR457396.1 | CAG33677.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1124863 | Chr.22:40861696 - 40864596 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map