Assay Details
Target Gene Details
Entrez Gene ID: | 54 |
Gene Name: | acid phosphatase 5, tartrate resistant |
Gene Aliases: |
HPAP, TRACP5a, TRACP5b, TRAP, TrATPase |
Location: |
Chr.19:11574660-11579008 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACP5 | NM_001111034.2 | 6 | 1350 | NP_001104504.1 |
| NM_001111035.2 | 7 | 1392 | NP_001104505.1 | |
| NM_001111036.2 | 6 | 1319 | NP_001104506.1 | |
| NM_001322023.1 | 6 | 1325 | NP_001308952.1 | |
| NM_001611.4 | 5 | 1216 | NP_001602.1 | |
| XM_005259938.1 | 7 | 1470 | XP_005259995.1 | |
| XM_011528069.2 | 7 | 1472 | XP_011526371.1 | |
| AK225912.1 | 6 | 1179 | ||
| AK290717.1 | 5 | 1181 | ||
| BC025414.2 | 5 | 1154 | AAH25414.1 | |
| BC111014.1 | 7 | 1335 | AAI11015.1 | |
| J04430.1 | 5 | 1154 | AAA76849.1 | |
| X14618.1 | 5 | 1102 | CAA32771.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509721 | Chr.19:11525216 - 11691390 on Build GRCh38 | Insertion |
|
| nsv519802 | Chr.19:11236817 - 11578343 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map