Assay Details
Target Gene Details
Entrez Gene ID: | 337880 |
Gene Name: | keratin associated protein 11-1 |
Gene Aliases: |
HACL-1, HACL1, KAP11.1 |
Location: |
Chr.21:30880644-30881555 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRTAP11-1 | NM_175858.2 | 1 | 230 | NP_787054.1 |
| AB096963.1 | 1 | 208 | BAE46378.1 | |
| AJ457065.1 | 1 | 230 | CAD29721.1 | |
| BC130555.1 | 1 | 223 | ||
| BC130557.1 | 1 | 223 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834078 | Chr.21:30847656 - 31014506 on Build GRCh38 | Gain |
|
| nsv834077 | Chr.21:30758503 - 30921578 on Build GRCh38 | Gain |
|
| nsv518454 | Chr.21:30842336 - 30927564 on Build GRCh38 | Loss |
|
| nsv1057467 | Chr.21:30318003 - 30993921 on Build GRCh38 | Gain |
|
| esv3646842 | Chr.21:30849249 - 30930214 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79258920] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map