Assay Details
Target Gene Details
Entrez Gene ID: | 27289 |
Gene Name: | Rho family GTPase 1 |
Gene Aliases: |
ARHS, RHO6, RHOS |
Location: |
Chr.12:48857133-48865870 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RND1 | NM_014470.3 | NP_055285.1 | ||
| AB040147.1 | BAB17851.1 | |||
| AF498967.1 | AAM21114.1 | |||
| AK292762.1 | ||||
| BC026356.1 | AAH26356.1 | |||
| Y07923.1 | CAA69228.1 |
More Information
Set Membership: |
|

Genomic Map