Assay Details
Target Gene Details
Entrez Gene ID: | 1733 |
Gene Name: | deiodinase, iodothyronine type I |
Gene Aliases: |
5DI, TXDI1 |
Location: |
Chr.1:53894187-53911086 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIO1 | NM_000792.6 | 4 | 1494 | NP_000783.2 |
| NM_001039715.2 | 3 | 1350 | NP_001034804.1 | |
| NM_001039716.2 | 3 | 1294 | NP_001034805.1 | |
| NM_001324316.1 | 2 | 1150 | NP_001311245.1 | |
| NM_213593.4 | 4 | 1302 | NP_998758.1 | |
| NR_136692.1 | 4 | 1385 | ||
| NR_136693.1 | 5 | 1411 | ||
| AW294928.1 | 1 | 384 | ||
| BC017955.1 | 3 | 1293 | AAH17955.2 | |
| BE046205.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011262 | Chr.1:53542956 - 54106304 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116793360] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map