Assay Details
Target Gene Details
Entrez Gene ID: | 10184 |
Gene Name: | lipoma HMGIC fusion partner-like 2 |
Gene Aliases: |
- |
Location: |
Chr.5:78485215-78648895 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LHFPL2 | NM_005779.2 | 5 | 2719 | NP_005770.1 |
| XM_006714515.1 | 4 | 2406 | XP_006714578.1 | |
| AY309920.1 | 2 | 2118 | AAP74733.1 | |
| BC136335.1 | 2 | 2070 | ||
| BC136336.1 | 2 | 2070 | ||
| CR936660.1 | 3 | 2356 | ||
| D86961.1 | 2 | 1938 | BAA13197.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1021871 | Chr.5:78459932 - 78579464 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map