Assay Details
Target Gene Details
Entrez Gene ID: | 57182 |
Gene Name: | ankyrin repeat domain 50 |
Gene Aliases: |
- |
Location: |
Chr.4:124664048-124712732 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD50 | NM_001167882.1 | 3 | 2246 | NP_001161354.1 |
| NM_020337.2 | 4 | 3521 | NP_065070.1 | |
| XM_017008471.1 | 3 | 4254 | XP_016863960.1 | |
| AB033049.2 | 1 | 1465 | BAA86537.2 | |
| AK091516.1 | 1 | 1537 | ||
| AK127496.1 | 3 | 2372 | BAC87007.1 | |
| AK291068.1 | 4 | 3521 | ||
| AK295135.1 | 3 | 2246 | ||
| BC024725.1 | 1 | 545 | AAH24725.1 | |
| BX640674.1 | 3 | 2038 | CAE45806.1 | |
| KU178729.1 | 1 | 425 | ||
| KU178730.1 |
More Information
Set Membership: |
|

Genomic Map