Assay Details
Target Gene Details
Entrez Gene ID: | 79781 |
Gene Name: | IQ motif containing with AAA domain 1 |
Gene Aliases: |
4930465P12Rik, DRC11, IQCA |
Location: |
Chr.2:236324147-236507558 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IQCA1 | NM_001270584.1 | NP_001257513.1 | ||
| NM_001270585.1 | NP_001257514.1 | |||
| NM_024726.4 | NP_079002.3 | |||
| NR_073043.1 | ||||
| XM_011511864.1 | XP_011510166.1 | |||
| XM_011511865.2 | XP_011510167.1 | |||
| XM_011511866.1 | XP_011510168.1 | |||
| XM_017004960.1 | XP_016860449.1 | |||
| AK054711.1 | BAB70798.1 | |||
| AK294105.1 | ||||
| AK295566.1 | ||||
| BC028699.1 | AAH28699.1 | |||
| BC043504.1 | AAH43504.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997438 | Chr.2:236210757 - 237034862 on Build GRCh38 | Gain |
|
| esv3575342 | Chr.2:236235177 - 236495792 on Build GRCh38 | Gain |
|
| nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map