Assay Details
Target Gene Details
Entrez Gene ID: | 93 |
Gene Name: | activin A receptor type 2B |
Gene Aliases: |
ACTRIIB, ActR-IIB, HTX4 |
Location: |
Chr.3:38454299-38493142 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACVR2B | NM_001106.3 | 5 | 614 | NP_001097.2 |
| XM_005265583.3 | 5 | 3076 | XP_005265640.1 | |
| XM_017007514.1 | 5 | 3055 | XP_016863003.1 | |
| XM_017007515.1 | 5 | 700 | XP_016863004.1 | |
| XM_017007516.1 | 5 | 4024 | XP_016863005.1 | |
| AB209449.1 | 4 | 4379 | BAD92686.1 | |
| BC096243.1 | 5 | 594 | AAH96243.1 | |
| BC096244.1 | 5 | 594 | AAH96244.1 | |
| BC096245.1 | 4 | 676 | AAH96245.1 | |
| BC099642.4 | 5 | 594 | AAH99642.1 | |
| BP352716.1 | ||||
| X77533.1 | 5 | 594 | CAA54671.1 |
More Information
Set Membership: |
|

Genomic Map