Assay Details
Target Gene Details
Entrez Gene ID: | 54960 |
Gene Name: | gem nuclear organelle associated protein 8 |
Gene Aliases: |
FAM51A1 |
Location: |
Chr.X:14002494-14030087 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GEMIN8 | NM_001042479.1 | 5 | 2603 | NP_001035944.1 |
| NM_001042480.1 | 4 | 2521 | NP_001035945.1 | |
| NM_017856.2 | 5 | 2691 | NP_060326.1 | |
| XM_017029617.1 | 5 | 1692 | XP_016885106.1 | |
| XM_017029618.1 | 5 | 1664 | XP_016885107.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528634 | Chr.X:13696631 - 14038859 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map