Assay Details
Target Gene Details
Entrez Gene ID: | 5859 |
Gene Name: | glutaminyl-tRNA synthetase |
Gene Aliases: |
GLNRS, MSCCA, PRO2195 |
Location: |
Chr.3:49095932-49105129 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| QARS | NM_001272073.1 | NP_001259002.1 | ||
| NM_005051.2 | NP_005042.1 | |||
| NR_073590.1 | ||||
| XM_017006965.1 | XP_016862454.1 | |||
| AF130067.1 | AAG35495.1 | |||
| AK056815.1 | ||||
| AK222510.1 | BAD96230.1 | |||
| AK290523.1 | ||||
| AK293260.1 | ||||
| AK297987.1 | ||||
| AK300220.1 | ||||
| AK301559.1 | ||||
| AK302867.1 | ||||
| BC000394.2 | AAH00394.1 | |||
| BC001567.1 | AAH01567.1 | |||
| BC001772.1 | AAH01772.2 | |||
| BC016634.1 | AAH16634.2 | |||
| BC029739.1 | AAH29739.1 | |||
| X76013.1 | CAA53600.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834686 | Chr.3:49068614 - 49233123 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map