Assay Details
Target Gene Details
Entrez Gene ID: | 705 |
Gene Name: | bystin like |
Gene Aliases: |
BYSTIN |
Location: |
Chr.6:41921227-41933046 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BYSL | NM_004053.3 | 7 | 1638 | NP_004044.3 |
| AK314147.1 | ||||
| BC007340.2 | 7 | 1374 | AAH07340.3 | |
| BC050645.1 | 7 | 1638 | AAH50645.1 | |
| BC062627.1 | 7 | 1609 | AAH62627.2 | |
| L36720.2 | 6 | 918 | AAC16603.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830646 | Chr.6:41916050 - 42096473 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map