Assay Details
Target Gene Details
Entrez Gene ID: | 5739 |
Gene Name: | prostaglandin I2 (prostacyclin) receptor (IP) |
Gene Aliases: |
IP, PRIPR |
Location: |
Chr.19:46610040-46625118 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGIR | NM_000960.3 | 3 | 1578 | NP_000951.1 |
| XM_005259093.2 | 3 | 2308 | XP_005259150.1 | |
| XM_005259095.3 | 3 | 1561 | XP_005259152.1 | |
| BC075814.1 | 3 | 1555 | AAH75814.1 | |
| BC110342.1 | 3 | 1541 | AAI10343.1 | |
| BQ185457.1 | ||||
| D25418.1 | 3 | 1556 | BAA05008.1 | |
| D29634.1 | 3 | 1531 | BAA06110.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833849 | Chr.19:46558831 - 46703060 on Build GRCh38 | Loss |
|
| nsv953589 | Chr.19:46601544 - 46636643 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map