Assay Details
Target Gene Details
Entrez Gene ID: | 7011 |
Gene Name: | telomerase associated protein 1 |
Gene Aliases: |
TLP1, TP1, TROVE1, VAULT2, p240 |
Location: |
Chr.14:20365667-20413540 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 47 - Exon 47 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TEP1 | NM_001319035.1 | NP_001305964.1 | ||
| NM_007110.4 | NP_009041.2 | |||
| XM_005268027.4 | XP_005268084.1 | |||
| XM_011537110.2 | XP_011535412.1 | |||
| AB209669.1 | BAD92906.1 | |||
| AK303307.1 | ||||
| BC126107.1 | ||||
| U86136.2 | AAC51107.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952812 | Chr.14:20371342 - 20387141 on Build GRCh38 | Deletion |
|
| esv23438 | Chr.14:20332606 - 20789719 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map