Assay Details
Target Gene Details
Entrez Gene ID: | 80013 |
Gene Name: | family with sequence similarity 188 member A |
Gene Aliases: |
C10orf97, CARP, DERP5, MST126, MSTP126, my042 |
Location: |
Chr.10:15778169-15860533 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM188A | NM_001318330.1 | 14 | 2045 | NP_001305259.1 |
| NM_024948.3 | 15 | 2097 | NP_079224.1 | |
| XM_005252600.2 | 14 | 1991 | XP_005252657.1 | |
| XM_005252602.2 | 13 | 1939 | XP_005252659.1 | |
| XM_006717508.2 | 14 | 1993 | XP_006717571.1 | |
| XM_011519693.1 | 15 | 2083 | XP_011517995.1 | |
| XM_011519694.1 | 16 | 2138 | XP_011517996.1 | |
| XM_017016672.1 | 17 | 2230 | XP_016872161.1 | |
| XM_017016673.1 | 15 | 2100 | XP_016872162.1 | |
| XM_017016674.1 | 13 | 1941 | XP_016872163.1 | |
| XM_017016675.1 | 13 | 1910 | XP_016872164.1 | |
| XM_017016676.1 | 12 | 1858 | XP_016872165.1 | |
| AF063600.1 | 10 | 1363 | AAG43159.1 | |
| AF176916.1 | 14 | 2007 | AAQ13660.1 | |
| AI573091.1 | 1 | 323 | ||
| AK023459.1 | 15 | 2084 | BAB14582.1 | |
| AL833873.1 | 15 | 2059 | CAD38730.1 | |
| BC067799.1 | 15 | 2083 | AAH67799.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831795 | Chr.10:15683863 - 15832969 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map