Assay Details
Target Gene Details
Entrez Gene ID: | 139189 |
Gene Name: | diacylglycerol kinase kappa |
Gene Aliases: |
- |
Location: |
Chr.X:50365407-50470850 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DGKK | NM_001013742.3 | NP_001013764.1 | ||
| XM_017029268.1 | XP_016884757.1 | |||
| AB183864.1 | BAD86792.1 | |||
| BC137319.1 | ||||
| BC137320.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508771 | Chr.X:50319296 - 50425828 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map