Assay Details
Target Gene Details
Entrez Gene ID: | 121364 |
Gene Name: | olfactory receptor family 10 subfamily A member 7 |
Gene Aliases: |
OR12-6 |
Location: |
Chr.12:55221025-55221975 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR10A7 | NM_001005280.1 | 1 | 608 | NP_001005280.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv559001 | Chr.12:55113681 - 55663924 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map