Assay Details
Target Gene Details
Entrez Gene ID: | 11322 |
Gene Name: | transmembrane channel like 6 |
Gene Aliases: |
EV1, EVER1, EVIN1, LAK-4P |
Location: |
Chr.17:78108775-78132427 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMC6 | NM_001127198.2 | 20 | 2776 | NP_001120670.1 |
| NM_001321185.1 | 20 | 2729 | NP_001308114.1 | |
| NM_007267.7 | 20 | 2680 | NP_009198.4 | |
| XM_005256995.1 | 20 | 2805 | XP_005257052.1 | |
| XM_011524255.1 | 19 | 2707 | XP_011522557.1 | |
| XM_011524256.1 | 19 | 2581 | XP_011522558.1 | |
| XM_011524257.2 | 14 | 1825 | XP_011522559.1 | |
| XM_011524258.1 | 11 | 1657 | XP_011522560.1 | |
| XM_017024107.1 | 19 | 2574 | XP_016879596.1 | |
| XM_017024108.1 | 19 | 2527 | XP_016879597.1 | |
| XM_017024109.1 | 18 | 2496 | XP_016879598.1 | |
| AB002405.2 | 12 | 1488 | BAA24179.2 | |
| AK074065.1 | 20 | 2679 | ||
| AK310734.1 | ||||
| AY099356.1 | 21 | 2678 | AAM44452.1 | |
| AY099357.1 | 13 | 1625 | AAM44453.1 | |
| AY236496.1 | 19 | 2645 | AAP69874.1 | |
| BC018346.1 | 13 | 1644 | AAH18346.2 | |
| BC023597.2 | 20 | 2660 | AAH23597.1 | |
| BC035648.1 | 13 | 1700 | AAH35648.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833554 | Chr.17:77977426 - 78151914 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map