Assay Details
Target Gene Details
Entrez Gene ID: | 79847 |
Gene Name: | major facilitator superfamily domain containing 13A |
Gene Aliases: |
C10orf77, TMEM180, bA18I14.8 |
Location: |
Chr.10:102460896-102480118 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFSD13A | NM_024789.3 | 10 | 1733 | NP_079065.2 |
| XM_006717973.3 | 12 | 1936 | XP_006718036.1 | |
| XM_006717974.3 | 9 | 1672 | XP_006718037.1 | |
| XM_011540160.2 | 11 | 2356 | XP_011538462.1 | |
| XM_011540163.2 | 11 | 1846 | XP_011538465.1 | |
| XM_011540167.2 | 8 | 1159 | XP_011538469.1 | |
| XM_011540169.2 | XP_011538471.1 | |||
| XM_017016654.1 | 11 | 1890 | XP_016872143.1 | |
| XM_017016655.1 | 10 | 1934 | XP_016872144.1 | |
| XM_017016656.1 | 11 | 1996 | XP_016872145.1 | |
| XM_017016657.1 | XP_016872146.1 | |||
| XM_017016658.1 | XP_016872147.1 | |||
| XM_017016659.1 | 10 | 1903 | XP_016872148.1 | |
| AK026182.1 | 6 | 939 | BAB15385.1 | |
| AK296583.1 | 10 | 1594 | ||
| AK301612.1 | ||||
| BC057787.1 | 6 | 1107 | AAH57787.2 | |
| BC067530.1 | 5 | 721 | ||
| BC067531.1 | 5 | 721 | ||
| BC113589.1 | 8 | 1548 |

Genomic Map